Orders LinkContactUsLinkComponent
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Style AA rubber coated, Stainless steel, anti-magnetic, anti-acid, 130 mm, Straight, sharp, fine
Catalog # 76376-238
Supplier #:  78325-AA
ESD Safe Tweezers, Electron Microscopy Sciences
Catalog # 76376-238
ESD Safe Tweezers, Electron Microscopy Sciences
ESD Safe Tweezers, Electron Microscopy Sciences
Style AA rubber coated, Stainless steel, anti-magnetic, anti-acid, 130 mm, Straight, sharp, fine
Price
$68.01
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Soft ESD safe cushion grips (resistively 10E8 ohm), these tweezers are ergonomically designed.
Specifications
  • Description:
    Style AA rubber coated
  • Material:
    Stainless steel, anti-magnetic, anti-acid
  • Tip style:
    Straight, sharp, fine
  • Overall length:
    130 mm