Orders LinkContactUsLinkComponent
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Ergonomic forceps, 140 mm, 1.5 mm, Needle serrated tips
Catalog # 76376-028
Supplier #:  77937-60
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Catalog # 76376-028
Ergonomic Paddle Handle Forceps, Electron Microscopy Sciences
Ergonomic forceps, 140 mm, 1.5 mm, Needle serrated tips
Price
$300.20
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Conventional forceps made of premium grade, hardened stainless steel with a protective coating for extra durability and protection from rust and corrosion.
  • Satin-matte finish reduces eye-fatiguing glare
  • Available in a variety of length, tip, and handle styles
Curved styles are well suited for retreiving specimens from the inside perimeter of transport jars and tissue separation in the water bath.
Specifications
  • Description:
    Ergonomic forceps
  • Tip configuration:
    Needle serrated tips
  • Overall length:
    140 mm
  • Tip length:
    1.5 mm