Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® 20 mm generator, PT-DA 20/2WEC-E192, 10 - 2500 ml, 20×192 mm, PT 2500 E
  76360-444
 :   Kinematica
 :  11030276
Polytron® Generator, PT-DA/2WEC-E192, Kinematica
  76360-444
Polytron® Generator, PT-DA/2WEC-E192, Kinematica
Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® 20 mm generator, PT-DA 20/2WEC-E192, 10 - 2500 ml, 20×192 mm, PT 2500 E

 
$3,752.79
   
Each
 

W-design for fibrous/stringy materials. Special design to prevent the teeth clogging. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.

  • Dispersing fibrous, stringy or solid samples (e.g., pieces of meat)
  • Standard dispersing, emulsifying and suspending tasks
 
  • Description:
    Polytron® 20 mm generator, PT-DA 20/2WEC-E192
  • Diameter ×L:
    20×192 mm
  • For use with:
    PT 2500 E
  • Sample volume:
    10 - 2500 ml