Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® 25 mm generator, PT-DA 25/2WEC-E192, 100 - 2500 ml, 25×192 mm, PT 2500 E
Catalog # 76360-438
Supplier:   Kinematica
Supplier #:  11030260
Polytron® Generator, PT-DA/2WEC-E192, Kinematica
Catalog # 76360-438
Supplier:  Kinematica
Polytron® Generator, PT-DA/2WEC-E192, Kinematica
Polytron® Generator, PT-DA/2WEC-E192, Kinematica

Polytron® 25 mm generator, PT-DA 25/2WEC-E192, 100 - 2500 ml, 25×192 mm, PT 2500 E

Price
$4,039.07
Log in for availability
Each
Product Details & Documents

W-design for fibrous/stringy materials. Special design to prevent the teeth clogging. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.

  • Dispersing fibrous, stringy or solid samples (e.g., pieces of meat)
  • Standard dispersing, emulsifying and suspending tasks
Specifications
  • Description:
    Polytron® 25 mm generator, PT-DA 25/2WEC-E192
  • Diameter ×L:
    25×192 mm
  • For use with:
    PT 2500 E
  • Sample volume:
    100 - 2500 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.