Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica

Polytron® 20 mm generator, PT-DA 20/BEC-E192 (BIOTRONA), 100 - 2000 ml, 20×192 mm, PT 2500 E
  76360-432
 :   Kinematica
 :  11030288
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica
  76360-432
Polytron® Generator,  PT-DA/BEC (BIOTRONA), Kinematica
Polytron® Generator, PT-DA/BEC (BIOTRONA), Kinematica

Polytron® 20 mm generator, PT-DA 20/BEC-E192 (BIOTRONA), 100 - 2000 ml, 20×192 mm, PT 2500 E

 
$3,155.81
   
Each
 

BIOTRONA® design high turbulence mixing. High-turbulence mixing head with low shear forces and energy inputs. Time saving with improved results compared to stirrers and dissolvers of up to 90%. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.

  • Fast dissolving and suspending of solid particles into liquids
  • Ideal for higher viscosities
  • Lower shear forces
 
  • Description:
    Polytron® 20 mm generator, PT-DA 20/BEC-E192 (BIOTRONA)
  • Diameter ×L:
    20×192 mm
  • For use with:
    PT 2500 E
  • Sample volume:
    100 - 2000 ml