Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Polytron® Generator, PT-DA/2EC-E192, Kinematica

Polytron® Generator, PT-DA/2EC-E192, Kinematica

Polytron® 20 mm generator, PT-DA 20/2EC-E192, 100 - 2000 ml, 20×192 mm, PT 2500 E
Catalog # 76360-418
Supplier:   Kinematica
Supplier #:  11030069
Polytron® Generator, PT-DA/2EC-E192, Kinematica
Catalog # 76360-418
Supplier:  Kinematica
Polytron® Generator, PT-DA/2EC-E192, Kinematica
Polytron® Generator, PT-DA/2EC-E192, Kinematica

Polytron® 20 mm generator, PT-DA 20/2EC-E192, 100 - 2000 ml, 20×192 mm, PT 2500 E

Price
$2,923.89
Log in for availability
Each
Product Details & Documents

Standard dispersing generators come with the EC (EasyClean) design and rotor/stator geometries with two, four or six teeth rows. This is all you need for popular applications in the preparation of samples, formulation development and small-scale production. Dispersing generators are made from high-alloy stainless steel 316L, electropolished, Ra≤1.6 μm as standard.

  • All standard dispersing, emulsifying and suspending tasks
  • Dispersing of plant, human or animal tissues, resins and pigments in a variety of fluids and volumes
  • Sample preparation for extraction and dissolution of organic materials
  • Intensive mixing, dissolving solids
Specifications
  • Description:
    Polytron® 20 mm generator, PT-DA 20/2EC-E192
  • Diameter ×L:
    20×192 mm
  • For use with:
    PT 2500 E
  • Sample volume:
    100 - 2000 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.