Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Linearity Thyroid, AUDIT Microcontrols

Linearity Thyroid, AUDIT Microcontrols

Linearity FLQ Thyroid for Roche Systems, Frozen liquid, Total T3, Total T4, TSH, Free T3, Free T4, 7 days when stored at 2...8 °C, 10×2 ml
Catalog # 76359-740
Supplier:   Audit Microcontrols
Supplier #:  K926M-5
Linearity Thyroid, AUDIT Microcontrols
Catalog # 76359-740
CAS Number:  
Linearity Thyroid, AUDIT Microcontrols
Linearity Thyroid, AUDIT Microcontrols

Linearity FLQ Thyroid for Roche Systems, Frozen liquid, Total T3, Total T4, TSH, Free T3, Free T4, 7 days when stored at 2...8 °C, 10×2 ml

CAS Number:  
Price
$352.67
Log in for availability
Kit of 5 (2ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Linearity FD Thyroid for Abbott Architect I Series, Linearity FLQ Thyroid for Roche systems and Linearity FD Thyroid for Siemens Centaur are assayed quality control materials consisting of five levels.

Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.

Specifications

Specifications

  • Description:
    Linearity FLQ Thyroid for Roche Systems
  • Size:
    10×2 ml
  • Stability:
    7 days when stored at 2...8 °C
  • Analyte:
    Total T3, Total T4, TSH, Free T3, Free T4
  • Type:
    Frozen liquid

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.