Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Linearity BNP, AUDIT Microcontrols

Linearity BNP, AUDIT Microcontrols

Linearity FD BNP for RAMP, Freeze dried, NT-proBNP, 3 days when stored at 2...8 °C, 5×1 ml
Catalog # 76359-724
Linearity BNP, AUDIT Microcontrols
Catalog # 76359-724
Supplier:  
CAS Number:  
Linearity BNP, AUDIT Microcontrols
Linearity BNP, AUDIT Microcontrols

Linearity FD BNP for RAMP, Freeze dried, NT-proBNP, 3 days when stored at 2...8 °C, 5×1 ml

CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Universal linearity BNP High, low and extended and linearity lipids for Siemens Centaur, linearity BNP for Siemens Stratus are assayed quality control materials consisting of five levels of human plasma.

Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.

Specifications

Specifications

  • Description:
    Linearity FD BNP for RAMP
  • Size:
    5×1 ml
  • Stability:
    3 days when stored at 2...8 °C
  • Analyte:
    NT-proBNP
  • Type:
    Freeze dried

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.