Linearity Immunoassay, AUDIT Microcontrols
Linearity FD immunoassay and Linearity FD Immunoassay for Abbott Architect I series are assayed quality control materials consisting of five levels.
Each kit has five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.
- Description:Linearity FD immunoassay for Abbott Architect I Series
- Size:5×5 ml
- Stability:5 days when stored at 2...8 °C
- Analyte:CEA, Ferritin, Folate, FSH, LH, Progesterone, Prolactin, Free PSA, Total PSA, Testosterone, TSH, Vitamin B12
- Type:Freeze dried
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



