Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Electrode Fill Solutions

Electrode Fill Solutions

3M KCl, Bottle, 25 ml, Barium, fluoride, calcium, lithium
Catalog # 76359-848
Supplier:   Mettler Toledo
Supplier #:  51343180
Electrode Fill Solutions
Catalog # 76359-848
Supplier:  Mettler Toledo
CAS Number:  
Electrode Fill Solutions
Electrode Fill Solutions

3M KCl, Bottle, 25 ml, Barium, fluoride, calcium, lithium

CAS Number:  
Price
$26.80
Log in for availability
Each
Product Details & Documents

FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.

Delivery information: Supplied in 25 ml (0.9 oz.) bottles.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    3M KCl
  • Ion:
    Barium, fluoride, calcium, lithium
  • Volume:
    25 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.