Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Electrolyte Solution

Electrolyte Solution

Electrolyte 1 M LiCl/EtOH, 6×30 ml, Bottle, pH, —
Catalog # 76359-846
Supplier #:  51350088
Electrolyte Solution
Catalog # 76359-846
CAS Number:  
Electrolyte Solution
Electrolyte Solution

Electrolyte 1 M LiCl/EtOH, 6×30 ml, Bottle, pH, —

CAS Number:  
Price
$98.90
Log in for availability
Pack of 6
Product Details & Documents

pH electrolyte and maintenance solution.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Electrolyte 1 M LiCl/EtOH
  • Size:
    6×30 ml
  • For:
    pH
  • Ion:

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.