Conductivity Standards, Sachets
250 ml, 84 μS/cm, Bottles
: 51302153
NIST-traceable standard for conductivity electrode calibration and verification.
- NIST-traceable
- Ensure freshness
- Improve data
Mettler Toledo sachets ensure a fresh portion of standard is used for each calibration, improving your conductivity and TDS data.
- Pack type:Bottles
- Size:250 ml
- Conductivity:84 μS/cm