Orders LinkContactUsLinkComponent
Conductivity Standards, Sachets
Conductivity Standards, Sachets
250 ml, 12.88 mS/cm, Bottles
Catalog # 76359-840
Supplier:   Mettler Toledo
Supplier #:  51350094
Conductivity Standards, Sachets
Catalog # 76359-840
Supplier:  Mettler Toledo
CAS Number:  
Conductivity Standards, Sachets
Conductivity Standards, Sachets
250 ml, 12.88 mS/cm, Bottles
CAS Number:  
Price
$32.90
Log in for availability
Each
Product Details & Documents
NIST-traceable standard for conductivity electrode calibration and verification.
  • NIST-traceable
  • Ensure freshness
  • Improve data
Mettler Toledo sachets ensure a fresh portion of standard is used for each calibration, improving your conductivity and TDS data.
Specifications
Specifications
  • Pack type:
    Bottles
  • Size:
    250 ml
  • Conductivity:
    12.88 mS/cm