Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Electrolyte Solution

Electrolyte Solution

Electrolyte 3 M/L KCl saturated AgCl, 250 ml, Bottle, pH, —
Catalog # 76359-832
Supplier:   Mettler Toledo
Supplier #:  51350074
Electrolyte Solution
Catalog # 76359-832
Supplier:  Mettler Toledo
CAS Number:  
Electrolyte Solution
Electrolyte Solution

Electrolyte 3 M/L KCl saturated AgCl, 250 ml, Bottle, pH, —

CAS Number:  
Price
$41.00
Log in for availability
Each
Product Details & Documents

pH electrolyte and maintenance solution.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    Electrolyte 3 M/L KCl saturated AgCl
  • Size:
    250 ml
  • For:
    pH
  • Ion:

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.