Quick-DNA/RNA™ Miniprep Plus Kits
Quick-DNA/RNA™ Miniprep Plus, for Cells, any tissues, blood, biological fluids, 10 Preps
Catalog # 76358-832
Supplier: Zymo Research
Supplier #: D7003T
Jump to:
Product Details & Documents
The Quick-DNA/RNA MiniPrep Plus kit combines Quick-DNA/RNA technology with the addition of DNA/RNA Shield, a preservation and lysis technology, and Proteinase K to enable easy, reliable, and rapid isolation from any biological sample including cells, solid tissue, and whole blood. The procedure uses Zymo-Spin column technology that results in high-quality genomic DNA and total RNA that is ready for any downstream application including reverse transcription, microarray, sequencing, etc.
Documents
Quick DNA RNA
Datasheet
Zymo-Research DNA Clean-Up & Gel DNA Recovery
Product Literature
Specifications
Specifications
- Description:Quick-DNA/RNA™ Miniprep Plus
- Size:10 Preps
- Details:for Cells, any tissues, blood, biological fluids
- Cat. No.:76358-832
- Binding Capacity:100 µg
- Yield:100 µg DNA and RNA (binding capacity), ≥50 µl (elution volume)