Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
PH PROBE INGOLD RANGE 0-12 12X225 MM
Catalog # 76358-090
Supplier:
Bellco Glass
Expand image
PH PROBE INGOLD RANGE 0-12 12X225 MM
Catalog # 76358-090
Supplier:
Bellco Glass
Supplier Number:
7803-21300