Order Entry
Puerto Rico
ContactUsLinkComponent
CELL SCREEN SS 150 MESH 94U 37 MM CS5
Catalog # 76356-934
Supplier:  Bellco Glass
undefined
CELL SCREEN SS 150 MESH 94U 37 MM CS5
Catalog # 76356-934
Supplier:  Bellco Glass
Supplier Number:  1985-00150