Order Entry
Puerto Rico
ContactUsLinkComponent
INSECT SYSTEM DIG MICRO-CARRIER 230V 3L
Catalog # 76356-648
Supplier:  Bellco Glass
undefined
INSECT SYSTEM DIG MICRO-CARRIER 230V 3L
Catalog # 76356-648
Supplier:  Bellco Glass
Supplier Number:  1965-70230