Order Entry
Puerto Rico
ContactUsLinkComponent
IOB W/MICRO-CARRIER FLASK 100MM 45MM 15L
Catalog # 76356-630
Supplier:  Bellco Glass
undefined
IOB W/MICRO-CARRIER FLASK 100MM 45MM 15L
Catalog # 76356-630
Supplier:  Bellco Glass
Supplier Number:  1965-61150