Order Entry
Puerto Rico
ContactUsLinkComponent
FLASK WATER JACKETED ONLY 100MM 8L 3/8IN
Catalog # 76356-590
Supplier:  Bellco Glass
undefined
FLASK WATER JACKETED ONLY 100MM 8L 3/8IN
Catalog # 76356-590
Supplier:  Bellco Glass
Supplier Number:  1965-58001