Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
TRIS-Buffered Saline, 10X Solution

TRIS-Buffered Saline, 10X Solution

TRIS-buffered saline, 10X solution, 4 L, 7.25 - 7.55
Catalog # 76348-706
Supplier:   RPI
Supplier #:  T60075-4000.0
TRIS-Buffered Saline, 10X Solution
Catalog # 76348-706
Supplier:  RPI
CAS Number:  
TRIS-Buffered Saline, 10X Solution
TRIS-Buffered Saline, 10X Solution

TRIS-buffered saline, 10X solution, 4 L, 7.25 - 7.55

CAS Number:  
Price
$155.08
Log in for availability
Each (4l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

TBS 10X Solution (Tris Buffered Saline 10X Solution) is a commonly used reagent in life science research. It is a buffered salt solution consisting of Tris (tris(hydroxymethyl)aminomethane) and sodium chloride (NaCl) dissolved in water. TBS 10X Solution is an essential reagent in life science laboratories for maintaining pH stability and providing appropriate ionic conditions for biological experiments.

Using TBS 10X Solution creates a stable pH environment for various biological experiments, such as protein and nucleic acid analysis. Tris serves as a buffering agent, helping to maintain a consistent pH level in the solution, while sodium chloride provides ionic strength to mimic physiological conditions. To dilute TBS 10X Solution to a lower concentration (e.g., 1X) - mix it with distilled water before using it an experiment. This diluted solution can be used for various applications including protein and nucleic acid blotting, immunohistochemistry, and molecular biology techniques.

Caution: Research or further manufacturing use only, not for food or drug use.

Specifications

Specifications

  • Description:
    TRIS-buffered saline, 10X solution
  • Size:
    4 L
  • pH:
    7.25 - 7.55
  • Cat. No.:
    76348-706
  • Concentration:
    10X
  • Appearance:
    Clear liquid
  • Storage Temperature:
    RT

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.