Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Tris acetate

Tris acetate

Plastic bottle, 500 g
Catalog # 76348-700
Irritant
Supplier:   RPI
Supplier #:  T60070-500.0
Tris acetate
Catalog # 76348-700
Supplier:  RPI
CAS Number:  6850-28-8
Tris acetate
Tris acetate

Plastic bottle, 500 g

CAS Number:  6850-28-8
Price
$321.84
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  Tris(hydroxymethyl)aminomethane acetate salt, 2-Amino-2-(hydroxymethyl)propane-1,3-diol acetate salt, Trimethylolaminomethane acetate, Tris acetate, Tris(hydroxymethyl)aminomethane acetate, 2-Amino-2-(hydroxymethyl)-1,3-propanediol acetate
  • CAS Number:6850-28-8
  • Formula:NH₂C(CH₂OH)₃·CH₃COOH
  • MDL Number:MFCD00038951
  • Molecular Weight:181.1870 g/mol
  • Storage Temperature:Room Temperature
  • EINECS:229-939-6
Specifications

Specification Test Results

  • Appearance:
    White crystalline powder
  • Heavy Metals:
    ≤0.001%
  • Moisture:
    ≤0.3%
  • pH (0.1M Solution in water):
    6.0 - 7.0
  • Purity:
    ≥98.0%
  • Residue on Ignition:
    ≤0.1%
  • BSE/TSE Free:
    BSE/TSE Free
  • Country of Origin:
    Report

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.