Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Mannitol Salt Agar

Mannitol Salt Agar

Mannitol salt agar, 500 g, 7.2 - 7.6
Catalog # 76346-916
Supplier:   RPI
Supplier #:  M43120-500.0
Mannitol Salt Agar
Catalog # 76346-916
Supplier:  RPI
CAS Number:  
Mannitol Salt Agar
Mannitol Salt Agar

Mannitol salt agar, 500 g, 7.2 - 7.6

CAS Number:  
Price
$127.31
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Caution: Research or further manufacturing use only, not for food or drug use.

Specifications

Specifications

  • Description:
    Mannitol salt agar
  • Size:
    500 g
  • pH:
    7.2 - 7.6

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.