Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Malt Extract Agar

Malt Extract Agar

500 g, At 25 °C: 4.6 ±0.2, Plastic bottle, RT
Catalog # 76346-776
Supplier:   RPI
Supplier #:  M21500-500.0
Malt Extract Agar
Catalog # 76346-776
Supplier:  RPI
CAS Number:  
Malt Extract Agar
Malt Extract Agar

500 g, At 25 °C: 4.6 ±0.2, Plastic bottle, RT

CAS Number:  
Price
$177.98
Log in for availability
Each (500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Malt Extract Agar is used for the isolation, cultivation and maintenance of yeasts and molds. Formulation is similar to Malt Extract Broth with the addition of agar and glycerol. This broth medium was recommended by Thom and Church for the cultivation of fungi. Its low acidity inhibits the growth of most bacteria. The high carbohydrate content enhances the rapid growth of fungi and yeasts.

Caution: Research or further manufacturing use only, not for food or drug use.

Documents

Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    500 g
  • pH:
    At 25 °C: 4.6 ±0.2
  • Storage temperature:
    RT
  • Cat. No.:
    76346-776
  • Solubility:
    Water with heat
  • CAS No.:
    8002-48-0

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.