Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acrylamide solution 40% (w/v)

Acrylamide solution 40% (w/v)

Plastic bottle, 5 L
Catalog # 76344-538
Toxic
Health
Supplier:   RPI
Supplier #:  A11265-5000.0
Acrylamide solution 40% (w/v)
Catalog # 76344-538
Supplier:  RPI
CAS Number:  79-06-1
Acrylamide solution 40% (w/v)
Acrylamide solution 40% (w/v)

Plastic bottle, 5 L

CAS Number:  79-06-1
Price
$1,164.51
Log in for availability
Each (5l)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

A 40% (w/v) Acrylamide solution.

Caution: Research or further manufacturing use only, not for food or drug use.

Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Storage Temperature:Refrigerator
  • UN:3426
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    Clear, Colorless Solution
  • Acrylic Acid:
    ≤ 0.003 %
  • Conductivity:
    ≤ 5 µmhos
  • Electrophoresis:
    Pass Test
  • pH @25 °C:
    6.0 - 7.0

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    5 L

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.