TUBE K2EDTA MINICOLLECT 0.25/0.5ML PK500
Catalog # 76343-512
Supplier: Greiner Bio-One
TUBE K2EDTA MINICOLLECT 0.25/0.5ML PK500
Catalog # 76343-512
Supplier: Greiner Bio-One
Supplier Number: 450532
Frequently Bought Together
Fold and Snap Tube Rack, 13 mm, 90-Place
Supplier: VWR International
VWR® Cleanroom Spiral Notebooks, Latex-Free
Supplier: VWR International
SCALPEL SAFETY BARD-PARKER SZ10 PK10
Supplier: Aspen Surgical Products
Monoject™ Blood Collection Tubes, Covidien
Supplier: Cardinal Health
Microvette® 300 Capillary Blood Collection Tubes with Collection Rim
Supplier: SARSTEDT INC
ANTIBODY SALL1 RABBIT PAB 50UL
Supplier: Biorbyt
Formalin prefilled 10%
Supplier: Mortech Manufacturing
TAG EAR 501-1000 APPLICATOR MONEL 0.8CM
Supplier: FINE SCIENCE TOOLS INC MS
L-Kynurenine
Supplier: Thermo Scientific Chemicals
VWR® B2T-Series Balances with Calibration Certificate
Supplier: VWR International
U-tek® Gel Pack Mats and Single Gel Pack Ice Packs, Sonoco ThermoSafe
Supplier: Sonoco Thermosafe
4-Aminoimidazole-5-carboxamide 95%
Supplier: Thermo Scientific Chemicals
pGL4.73[hRluc/SV40] Vector, 20 µg, Promega
Supplier: Promega Corporation
Spectrophotometer Cuvettes, Chemglass
Supplier: Chemglass
PRECISION TROCHAR 10 GAUGE
Supplier: INNOVATIVE RESEARCH OF AM MS
Supreme Mini-Treats™, Certified, Bio-Serv
Supplier: Bio-Serv
FAP HIS TAG HUMAN PE-LABELED 25TESTS
Supplier: ACROBIOSYSTEMS INC MS
Anti-CD4 Rat Monoclonal Antibody [clone: GK1.5]
Supplier: Bio X Cell