Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
JACKET BOMBER PLUS 2-1 PU/CTN CL3 OR X3
Catalog # 76342-074
Supplier:
MCR Safety
Expand image
JACKET BOMBER PLUS 2-1 PU/CTN CL3 OR X3
Catalog # 76342-074
Supplier:
MCR Safety
Supplier Number:
BP2CL3OX3