RECOM PD-L1/B7-H1 HUMAN CF >95% 50UG
Catalog # 103781-762
Supplier: R&D Systems
RECOM PD-L1/B7-H1 HUMAN CF >95% 50UG
Catalog # 103781-762
Supplier: R&D Systems
Supplier Number: BT156-050
Frequently Bought Together
Static Mouse Cage, 1284L
Supplier: Tecniplast Usa
Reusable Plastic Animal Cages, Tecniplast®
Supplier: Tecniplast Usa
PVC Film Wrap, T FRANK MCCALLS INC
Supplier: TFrank McCalls
VWR® Shelf Liners
Supplier: VWR International
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
Pierce™ Recombinant C-Myc Peptide
Supplier: Invitrogen
KIT CUSTOM CUT 2-LAYER DRAWER LINER FOAM
Supplier: GLOBAL EQUIPMENT MS
VECTOR 8E+12VG 2E+13VG/ML CONC 4X0.1ML
Supplier: VIROVEK, INC. MS
SP Bel-Art Fritware® Porous Polyethylene Sheets, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
COVER SHOE STANDARD SZ 12-15 CS150
Supplier: ULINE MS
RECOM HUMAN PD-L2 FC BIOTINYLATED 50UG
Supplier: R&D Systems
FILTER PAPER BENCHKOTE 495 X 457MM PK50
Supplier: Cytiva
Nalgene® Polyolefin Plastic Paper Sheets, Thermo Scientific
Supplier: Thermo Fisher Scientific
PADS LIGHT DUTY CLEANING SYSTEM 10IN
Supplier: ULINE MS
UNDERPAD HIGH-ABSORBANCE 23X36IN CS72
Supplier: COVETRUS NORTH AMERICA MS
Swab, Foam Tip, Sterile
Supplier: Hardy Diagnostics
neodisher® Detergents, Miele
Supplier: Miele