Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical

Empore™ Solid Phase Extraction RAD Disks, CDS Analytical

Empore™ RAD disk, Strontium (Sr-90), 47 mm, 0.5 mm
Catalog # 76333-106
Supplier:   CDS ANALYTICAL
Supplier #:  98-0405-0064-1
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Catalog # 76333-106
Supplier:  CDS ANALYTICAL
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical
Empore™ Solid Phase Extraction RAD Disks, CDS Analytical

Empore™ RAD disk, Strontium (Sr-90), 47 mm, 0.5 mm

Price
$2,962.56
Log in for availability
Pack of 20 Case of 60
Product Details & Documents

Empore RAD disks provide an efficient alternative to conventional radiochemical sample preparation methods that use wet chemistry or packed columns.

  • EPA compliant with radiochemical testing methods
  • Improved safety by reducing usage of concentrated acids needed for precipitation methods
  • Easier sample handling and waste management by avoiding excessively large sample volumes needed for other radiochemical testing methods
  • Large-volume processing – Ultra-fast flow up to 700 mL/min can be achieved
  • Improved extraction efficiency – High density sorbent bed improves interactions between sorbent particles and analyte
  • Excellent reproducibility – Uniform particle bed improves lot-to-lot consistency

Empore RAD SPE disks trap radiochemical species by molecular recognition technology. Concentration, separation, and sample counting preparation are all easily combined into a single step.

Specifications
  • Ø:
    47 mm
  • Description:
    Empore™ RAD disk, Strontium (Sr-90)
  • Thickness:
    0.5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.