RECOM PD-L1/B7-H1 HIS-TAG AVI-TAG 50 UG
Catalog # 103751-716
Supplier: R&D Systems
RECOM PD-L1/B7-H1 HIS-TAG AVI-TAG 50 UG
Catalog # 103751-716
Supplier: R&D Systems
Supplier Number: AVI9049-050
Frequently Bought Together
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Bead Ruptor Pre-Filled Bead Tubes, 2 ml, Omni International
Supplier: OMNI INTERNATIONAL
Corning® DMEM (Dulbecco's Modification of Eagle's Medium), Corning
Supplier: Corning
VWR® Universal Pipette Tips
Supplier: VWR International
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
Acetone ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International