Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER TBS BLOCKING READY-TO-USE 3X500ML
Catalog # 103749-018
Supplier:
Li-Cor
Expand image
BUFFER TBS BLOCKING READY-TO-USE 3X500ML
Catalog # 103749-018
Supplier:
Li-Cor
Supplier Number:
927-60003