Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CRUCIBLE 5 % GOLD 95 % PLATINUM STD 10ML
Catalog # 76336-716
Supplier:
XRF Scientific
Expand image
CRUCIBLE 5 % GOLD 95 % PLATINUM STD 10ML
Catalog # 76336-716
Supplier:
XRF Scientific
Supplier Number:
5AUPTCR10L