Order Entry
Puerto Rico
ContactUsLinkComponent
STAND VC 10.400 100 X 175 X 160 MM 16 MM
Catalog # 76329-792
Supplier:  IKA Works
undefined
STAND VC 10.400 100 X 175 X 160 MM 16 MM
Catalog # 76329-792
Supplier:  IKA Works
Supplier Number:  0035001735