ContactUsLinkComponent
COLUMN MP C18(2) 110A 2.1X100MM 5UM
Catalog # 76329-240
Supplier:  Phenomenex
COLUMN MP C18(2) 110A 2.1X100MM 5UM
Catalog # 76329-240
Supplier:  Phenomenex
Supplier Number:  VA951002-2
Frequently Bought Together