Basic IV Administration Sets, B. Braun
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Documents
Specifications
- Flow rate:15 Drops/ml
- Sterility:Sterile
- Description:Universal Spike, Roller Clamp, Slide Clamp and SPIN-LOCK® Connector, Priming Volume: 13 ml, Labeled 'Contents Sterile'
- Length:215.9 cm (85")