Orders LinkContactUsLinkComponent
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Universal Spike, Backcheck Valve, 0.2 µm Filter, 3 Injection Sites, Roller Clamp, Slide Clamp, SPIN-LOCK® Connector, Priming Volume: 20 ml, 15 Drops/ml, 211 cm (83"), Sterile
Catalog # 76327-242
Supplier:   B Braun
Supplier #:  352447
Basic IV Administration Sets, B. Braun
Catalog # 76327-242
Supplier:  B Braun
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Universal Spike, Backcheck Valve, 0.2 µm Filter, 3 Injection Sites, Roller Clamp, Slide Clamp, SPIN-LOCK® Connector, Priming Volume: 20 ml, 15 Drops/ml, 211 cm (83"), Sterile
Price
$1,026.85
Log in for availability
Case of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Specifications
  • Flow rate:
    15 Drops/ml
  • Sterility:
    Sterile
  • Description:
    Universal Spike, Backcheck Valve, 0.2 µm Filter, 3 Injection Sites, Roller Clamp, Slide Clamp, SPIN-LOCK® Connector, Priming Volume: 20 ml
  • Length:
    211 cm (83")