Orders LinkContactUsLinkComponent
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Rate Flow Regulator with Female Luer Connector, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 2.6 ml, 46 cm (18"), Non sterile
Catalog # 76327-092
Supplier #:  473007
Basic IV Administration Sets, B. Braun
Catalog # 76327-092
Supplier:  
Basic IV Administration Sets, B. Braun
Basic IV Administration Sets, B. Braun
Rate Flow Regulator with Female Luer Connector, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 2.6 ml, 46 cm (18"), Non sterile
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
B. Braun gravity sets are available in various lengths and dimensions to accommodate various applications.
Specifications
  • Sterility:
    Non sterile
  • Description:
    Rate Flow Regulator with Female Luer Connector, Injection Site 6" Above Distal End, SPIN-LOCK® Connector, Priming Volume: 2.6 ml
  • Length:
    46 cm (18")