$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
KIT 96-WELL BIO ASSAY NON-STERILE CS1
76324-018
:
Greiner Bio-One
KIT 96-WELL BIO ASSAY NON-STERILE CS1
76324-018
:
Greiner Bio-One
:
655846
Greiner Bio-One 3D cell culture
Greiner Bio-One Product Catalog
Frequently Bought Together
VWR® Disposable Hexagonal Weigh Boats
: VWR International
VWR® Disposable Serological Pipettes
: VWR International
Eppendorf® Safe-Lock Tubes, 2.0 ml - Microtubes
: Eppendorf
VWR® Ultra High Performance Centrifuge Tubes
: VWR International
Eppendorf® DNA LoBind® Microcentrifuge Tubes
: Eppendorf
VWR® Microcentrifuge Tubes, PP
: VWR International
VWR® High Performance Microcentrifuge Tubes, PP
: VWR International
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: Eppendorf
Eppendorf® Safe-Lock Tubes, 1.5 ml - Microtubes
: Eppendorf
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
: VWR International
VWR® Disposable Transfer Pipettes
: VWR International
VWR® Disposable Microcentrifuge Tubes
: VWR International
Eppendorf Tubes® Microtubes with Snap Cap, 5.0 ml
: Eppendorf
Axygen® Snaplock Microcentrifuge Tubes, 0.6/1.5/1.7/2.0/5.0 ml, Corning
: Corning