STANDARD SOLUTION 500MOSM/KG 125ML
Catalog # MSPP-2105
Supplier: PRECISION SYSTEMS INC MS
STANDARD SOLUTION 500MOSM/KG 125ML
Catalog # MSPP-2105
Supplier: PRECISION SYSTEMS INC MS
Supplier Number: 2105
Frequently Bought Together
TSK STERiJECT® Premium Needles, Air-Tite Products
Supplier: Air-Tite
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Universal Vial Caps, Ab Sciex
Supplier: AB SCIEX LLC MS
STANDARD SOLUTION 100MOSM/KG 125ML
Supplier: PRECISION SYSTEMS INC MS