STANDARD SOLUTION 100MOSM/KG 125ML
Catalog # MSPP-2101
Supplier: PRECISION SYSTEMS INC MS
STANDARD SOLUTION 100MOSM/KG 125ML
Catalog # MSPP-2101
Supplier: PRECISION SYSTEMS INC MS
Supplier Number: 2101
Frequently Bought Together
Ward's® Bulk Simulated Antisera
Supplier: Avantor
RECOM IFN-GAMMA MS PURITY>97% CF 100UG
Supplier: Novus Biologicals
ELISA KIT TAC1 IMMUNOTAG HU 96WELL PLATE
Supplier: G-Biosciences
High Sensitivity DNA ScreenTape Analysis
Supplier: Agilent Technologies
Static Rat/Hamster Cages, 2154F
Supplier: Tecniplast Usa
MAB MOUSE CD95 DX2 HU BV 711 100 TESTS
Supplier: Biolegend
Pierce™ Antibody Coated Plates, 96 Well,, Thermo Scientific
Supplier: Invitrogen
neodisher® Detergents, Miele
Supplier: Miele
pH Buffer Capsules, Hydrion®
Supplier: Micro Essential
Lithium carbonate ≥99.998% (metals basis excluding Ca), Puratronic® (max. 0.002% Ca)
Supplier: Thermo Scientific Chemicals
BUFFER NEUTRALIZATION NEU EF 1000 ML
Supplier: MACHEREY-NAGEL INC MS
AGAR BRUCELLA BLOOD 1 PLATE
Supplier: ANAEROBE SYSTEMS MS
STANDARD SOLUTION 500MOSM/KG 125ML
Supplier: PRECISION SYSTEMS INC MS
Nalgene® PolyPaper® Right-To-Know Custom Labeling System, Thermo Scientific
Supplier: Thermo Fisher Scientific
Molecular sieve 3A (0.3 nm, 3 Å), pellets 1/16''
Supplier: Strem Chemicals Inc