GLYCOCATCH RESIN FOR ENRICHMENT 200UG
Catalog # MSPP-G3OC6002
Supplier: GENOVIS, INC MS
GLYCOCATCH RESIN FOR ENRICHMENT 200UG
Catalog # MSPP-G3OC6002
Supplier: GENOVIS, INC MS
Supplier Number: G3-OC6-002
Frequently Bought Together
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Pharmalytes® IEF Carrier Ampholytes, Cytiva
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
PI MARKER MAURICE CIEF 4.05 210UL
Supplier: PROTEIN SIMPLE MS
MARKER MAURICE CIEF PI-8.40 LYOPH 210UL
Supplier: PROTEIN SIMPLE MS
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
DNA WASH BUFFER 6 ML
Supplier: Zymo Research
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
EMITTER NEWOMICS M3 20UM 8 NOZZLES
Supplier: NEWOMICS INC. MS
Unstained Protein Standard, Broad Range, New England Biolabs
Supplier: New England Biolabs (NEB)
Grade
Supplier: Promega Corporation
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
Blue Prestained Protein Standard, Broad Range (11 to 250 kDa), New England Biolabs
Supplier: New England Biolabs (NEB)
TEV PROTEASE HIS >95% 10000IU
Supplier: Genscript
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS