SODIUM HYDROXIDE SOLUTION 50% W/V 1L
Catalog # 103742-318
Supplier: Aqua Solutions
SODIUM HYDROXIDE SOLUTION 50% W/V 1L
Catalog # 103742-318
Supplier: Aqua Solutions
Supplier Number: 8619-1L
Frequently Bought Together
Parafilm® M Sealing Film
Supplier: Heathrow Scientific
VWR® Weighing Paper
Supplier: VWR International
VWR® Laboratory Bench and Table Protectors with Leakproof Moisture Barrier
Supplier: VWR International
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
VWR® Multipurpose Containers, HDPE
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
Nalgene® VERSI-DRY® Lab Table Soakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
Chemical Spill Tray
Supplier: SCIENTIFIC PLASTICS
NeoPro® Powder-Free Neoprene Gloves, Microflex®, Ansell
Supplier: ANSELL HEALTHCARE PRODUCTS LLC MX
Blood Agar Plate, 5% Sheep Blood in Tryptic Soy Agar (TSA) Base, 15×100 mm plate
Supplier: Hardy Diagnostics
Genomic DNA ScreenTape Analysis
Supplier: Agilent Technologies
Hexane (mixture of isomers) ACS, VWR Chemicals BDH®
Supplier: VWR International
VIAL INSERT FOR ICE280 AND ICE3 300 UL
Supplier: PROTEIN SIMPLE MS
RNA ScreenTape Analysis
Supplier: Agilent Technologies
Dichloromethane ≥99.5% stabilised ACS, VWR Chemicals BDH®
Supplier: VWR International
Ethyl acetate ≥99.5% ACS, VWR Chemicals BDH®
Supplier: VWR International
Ethanol 96% (v/v), EMPROVE® EXPERT Ph. Eur., BP, ChP, SAFC®
Supplier: MilliporeSigma
Criterion™ TGX™ Precast Gels, Midi Protein Gels, Bio-Rad
Supplier: Bio-Rad