Orders LinkContactUsLinkComponent
Axygen® MultiRack Tipstations, Corning
0.1 - 10 µl, Stack: 960 Tips/Stack, 5 Stacks/Case
  76320-706
 :   Corning
 :  MR-10XT-TS
Axygen® MultiRack Tipstations, Corning
  76320-706
 :  Corning
Axygen® MultiRack Tipstations, Corning
Axygen® MultiRack Tipstations, Corning
0.1 - 10 µl, Stack: 960 Tips/Stack, 5 Stacks/Case
$455.00
Case of 4,800
The Axygen® MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter and reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibition free
  • Non pyrogenic
  • Manufactured in an ISO certified facility in the USA
Axygen® MultiRack tipstation is packaged in a format with 192 tips per layer and five layers total and can be used with 8 or 12 channel pipettors. This packaging style provides a convenient solution for lab applications such as ELISA, PCR, and NGS library preparation, where thousands of tips are consumed. Axygen® MultiRack tipstation is available with 10 and 200 µl, sterile or non sterile.
 : Axygen® MultiRack pipette tips are also available in bulk, hinged rack, filter tips and reload configurations.
  • Volume:
    0.1 - 10 µl
  • Packed:
    Stack: 960 Tips/Stack, 5 Stacks/Case
Accessories
  •  
    Description
    For use with
  • 76320-746
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    200 µl extended length, 300 µl extended length, and 1000 µl extended length tips
    Case of 5 - $142.00
  •  
    Description
    For use with
  • 76320-744
    MultiRack Empty Rack, 5/Pack, 1 Pack/Case
    10 µl extended length, 200 µl, 200 µl NX, 300 µl tips
    Case of 5 - $141.00