Camera Adapters
C-mount adapter for 1/2" and 1/3" chip sensors, 0.50×, Compatible cameras: Moticam 3+, Moticam 5+, Moticam 10+, Moticam BTU/BTW series and connects with AE31E/AE2000, BA series and SMZ161/171 series
Catalog # 470314-902
Supplier: Motic
: 1101001901791
Jump to:
Product Details & Documents
C-mount camera adapter allows Moticams to connect with Motic trinocular microscopes.
Specifications
- Description:C-mount adapter for 1/2" and 1/3" chip sensors, 0.50×
- For use with:Compatible cameras: Moticam 3+, Moticam 5+, Moticam 10+, Moticam BTU/BTW series and connects with AE31E/AE2000, BA series and SMZ161/171 series