Biox Micro PettePlus Pipettes
MicroPette Plus Pipette, 10 - 100 µl, 1 µl
Supplier: TWD Scientific
Supplier #: BX01-2104
Jump to:
Product Details & Documents
MicroPette Plus mechanical pipettes are the ideal adjustable volume pipettes for all of your basic pipetting needs.
- Ergonomic design for comfortable extended use
- Easy-to-read display
- Convenient one-handed operation
- Easy to calibrate and maintain with included tool
- Individual piston and tip cone assemblies allow for easy repair and maintenance
- Fully autoclavable
- Compatible with most standard pipette tips
- 4 sizes available
Micropette plus features single-channel adjustable volume. Adjustable in various increments depending on size.
Certifications: The MicroPette Plus is calibrated in accordance with ISO 8655, and each pipette includes its own individual test certificate
Accessories information: Pipette Stand is available to accommodate up to 6 pipettes (01-2306)
Documents
Specifications
- Volume:10 - 100 µl
- Description:MicroPette Plus Pipette
- Increments:1 µl