Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Chorionic Gonadotropin Human

Chorionic Gonadotropin Human

5 mg
Catalog # 103737-128
Marketsource
Supplier:   BioVendor
Supplier #:  RP17825005
Chorionic Gonadotropin Human
Catalog # 103737-128
Supplier:  BioVendor
Marketsource
CAS Number:  
Chorionic Gonadotropin Human
Chorionic Gonadotropin Human

5 mg

CAS Number:  
Price
$733.69
Log in for availability
Each (5mg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human Chorionic Gonadotropin is produced from a sterile preparation of placental glucoprotein urine of pregnant women having a total molecular mass of 36700 Dalton.

  • Urine of pregnant women
  • Sterile Filtered White lyophilized (freeze-dried) powder
  • The hCG was lyophilized with no additives
  • It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm
  • H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions

The hCG consists of 237 amino acids, a chain-92 amino acids and b chain-145 amino acids. The hCG is purified by proprietary chromatographic techniques.

Caution: Research use only.

Specifications

Specifications

  • Size:
    5 mg
  • Storage Conditions:
    Lyophilized hCG although stable at room temperature for 3 weeks, should be stored desiccated below −18 °C. Upon reconstitution CG-beta should be stored at 4 °C between 2 - 7 days and for future use below −18 °C. For long term storage it is recommended to add a carrier protein (0.1% HSA or BSA). Please prevent freeze-thaw cycles
  • Cat. No.:
    103737-128
  • Biological Activity:
    The activity was found to be 5625 IU/mg (may vary slightly with each lot)
  • Protein/Peptide Type:
    Native
  • Formulation:
    The hCG was lyophilized with no additives
  • Reconstitution Instructions:
    It is recommended to reconstitute the lyophilized Human Chorionic Gonadotropin in sterile 18 MΩ-cm H₂O not less than 100 µg/ml, which can then be further diluted to other aqueous solutions.
  • Protein Synonyms:
    CGH
  • Protein/Peptide Name:
    Chorionic Gonadotropin Human
  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Source:
    Urine (pregnant women)

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.