Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REACTION TUBE PP NATURAL W/CAP 5ML PK200
Catalog # 76311-044
Supplier:
Greiner Bio-One
Expand image
REACTION TUBE PP NATURAL W/CAP 5ML PK200
Catalog # 76311-044
Supplier:
Greiner Bio-One
Supplier Number:
622201
Related Information
Documentation
Product Literature
Greiner Bio-One Product Catalog
Documentation