Objectives for REBEL Microscopes, ECHO
60× Plan Apochromat, CSC 0.17, 0.26 mm, N.A. 0.90
Catalog # 76307-588
Supplier: DISCOVER ECHO INC.
: RMAG9602
Jump to:
Product Details & Documents
Specifications
- Description:60× Plan Apochromat, CSC 0.17
- Aperture:N.A. 0.90
- Working distance:0.26 mm