Objectives for REBEL Microscopes, ECHO
4× Plan Apochromat, CSC 0.17, 16.6 mm, N.A. 0.13
Catalog # 76307-578
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG9042
Jump to:
Product Details & Documents
Specifications
- Description:4× Plan Apochromat, CSC 0.17
- Aperture:N.A. 0.13
- Working distance:16.6 mm