Objectives for REBEL Microscopes, ECHO
20× Plan Achromat Phase, CSC 0.17, 1.6 mm, N.A. 0.40
Catalog # 76307-566
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG0203
Jump to:
Product Details & Documents
Specifications
- Description:20× Plan Achromat Phase, CSC 0.17
- Aperture:N.A. 0.40
- Working distance:1.6 mm